missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ BHI Broth (Brain Heart Infusion Broth)
Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.
£21.41 - £125.00
Specifications
Certifications/Compliance | Industrial Reference: BAM, SMD, SMWW, USDA. |
---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
17742998
|
Thermo Scientific™
R060264 |
100 x 4 mL | 15mm x 103mm Tube | N/A | |||||
17752998
|
Thermo Scientific™
R060266 |
20 x 4 mL | 15mm x 103mm Tube | N/A | |||||
17762998
|
Thermo Scientific™
R060270 |
100 x 5 mL | 15mm x 103mm Tube | N/A | |||||
17772998
|
Thermo Scientific™
R060272 |
20 x 5 mL | 15mm x 103mm Tube | N/A | |||||
Description
Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.
Specifications
Industrial Reference: BAM, SMD, SMWW, USDA. |
REF R14101 for use with MicroScan™ System. REF R112022 not intended for IVD use.