Learn More
Thermo Scientific™ Coagulase Plasma
Detect coagulase enzyme in staphylococci with Coagulase Plasma.
Brand: Thermo Scientific™ R21052
92.63 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
Description
Detect coagulase enzyme in staphylococci with Thermo Scientific™ Remel™ Coagulase Plasma. The enzyme coagulase acts on a constituent of rabbit plasma to produce a thrombin-like substance which activates fibrinogen to form fibrin clot. In 1940, Fairbrother and Chapman et al. reported pathogenic staphylococci can be identified based on the ability to coagulate plasma.1,2 Chapman found rabbit plasma to be superior to other types of plasma in terms of clotting activity.3 Bayliss and Hall recommended replacing the citrate anticoagulant with EDTA to avoid false-positive clots.4
- Rabbit plasma with EDTA
- Lyophilized
Specifications
Coagulase Plasma | |
Staphylococcus species | |
Enzyme Immunoassay Detection |
Yes | |
Coagulase Plasma | |
25 mL |