Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated)
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
£143.00
Specifications
Description | Contact Plates; Sterile; Tryptic Soy Agar w/Lecithin; Gamma irradiated |
---|---|
Format | Contact - Double Bag |
Product Type | Agar |
Description
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
- Double-bagged plus a third bag is enclosed for customer use (Double + Bag)
- Products are manufactured in a clean room and validated sterile by gamma radiation
- Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifications
Contact Plates; Sterile; Tryptic Soy Agar w/Lecithin; Gamma irradiated | |
Contact - Double Bag | |
Agar |
Not intended for IVD use.