Learn More
Thermo Scientific™ Remel™ Gram Safranin
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
Brand:  Thermo Scientific™ R40059
 View more versions of this product
Product Code. 12623668
  • £151.00 / Pack of 5
Explore more special offers
Description
Description
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Safranin
5 x 250 mL
5/Pk.
CE Marked
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.