Learn More
Thermo Scientific™ Remel™ Isolator Wrap™ Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80
Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80 is used for the cultivation of yeasts and fungi.
Brand: Thermo Scientific™ R111825
Additional Details : Weight : 0.60000kg
Description
Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80 is used for the cultivation of yeasts and fungi.
- Foil Barrier Wrap — impermeable to isolator decontamination process
- GripFit™ Plates — easy to handle with one hand without lid separating from bottom of plate
- Irradiated — Sterility Assurance Level 10-6
- Shrink-wraped with easy open tear strip — reduces breakage and moisture loss
- Bottom labeling — information stays with sample
- Extra sterile bag — easy to transport plates to lab without introducing contamination
- Six month shelf life — less ordering and fewer lot numbers to QC
Specifications
Sabouraud Dextrose Agar | |
Foil Barrier Wrap + Bag | |
Agar | |
10/Pk. |
Not intended for IVD use.