missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Broth (Dehydrated)

Selectively and differentially isolate enteric gram-negative bacilli using MacConkey Broth.

Brand:  Thermo Scientific™ R453822

207.27 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12938244

  • £434.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Selectively and differentially isolate enteric gram-negative bacilli using Thermo Scientific™ Remel™ MacConkey Broth (Dehydrated). MacConkey Broth is a modification of the original bile salt broth first described by MacConkey in 1900, which contained litmus as an indicator and sodium taurocholate to inhibit gram-positive organisms1. In further testing, MacConkey recommended replacing litmus with neutral red2. In 1953, Childs and Allen replaced neutral red with bromocresol purple, a less inhibitory indicator3.

Selectively and differentially isolate enteric gram-negative bacilli using MacConkey Broth. This medium is used for the presumptive identification of coliforms. Coliforms are organisms that are present in the environment and in the feces of all warm-blooded animals and humans.

Their presence in drinking water indicates that disease-causing organisms (pathogens) could be in the water system.

  • Selective: Oxgall is a selective agent which inhibits gram-positive organisms.
  • Ease of Differentiation: Lactose and bromocresol purple indicator enable the differentiation of lactose fermenting gram-negative bacilli.
  • Conforms to USP and EP: The medium is formulated and tested in conformance with harmonized United States Pharmacopeia (USP) / European Pharmacopeia (EP) guidelines4,5 for the detection of Escherichia coli.

This medium contains gelatin peptone that provides amino acids and essential nutrients for bacterial growth. Lactose is a carbon source for energy. Oxgall is a selective agent which inhibits gram-positive organisms. Lactose and bromocresol purple pH indicator enable the differentiation of lactose fermenting gram-negative bacilli.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industrial Reference: USP
Powder
500 g
MacConkey Broth
Dehydrated Culture Media
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Instructions

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number165310 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog NumberR453822
Lot Number118371 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog NumberR453822
Lot Number835578 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR453822
Lot Number761564 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR453822
Lot Number698056 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR453822
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MacConkey Broth (Dehydrated) > MacConkey Broth; 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only