Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Brand: Thermo Scientific™ R20411
85.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
- Tube size is 15 x 103mm and fits in most spectrophotometers
- Polystyrene technology allows longer shelf life
Specifications
Turbidity Standard 1.0 | |
15 x 103 mm |