Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Brand:  Thermo Scientific™ R20413
85.87 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 13246239
  • £109.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbidity Standard 3.0
15 x 103 mm