missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Mono-Lex System
Rapid latex agglutination test used for detection of heterophile (Paul-Bunnell) antibodies associated with infectious mononucleosis in human serum or plasma.
£181.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Detectable Analytes | Mononucleosis Antibodies |
Sample Type | Plasma, Serum |
Type | Reagent |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12938818
|
Thermo Scientific™
R421021 |
100 Tests |
£181.00
100 tests |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific™ Remel Mono-Lex System is a rapid latex agglutination test used for detection of heterophile (Paul-Bunnell) antibodies associated with infectious mononucleosis in human serum or plasma.
Specifications
2°C to 8°C | |
Mononucleosis Antibodies | |
Plasma, Serum | |
Reagent |