missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel RSV Reagent
Remel PathoDx and Respiratory Virus Panel RSV Reagent is used with PathoDx Respiratory Virus Panel.
Brand: Thermo Scientific™ R62411
This item is not returnable.
View return policy
Description
Remel PathoDx and Respiratory Virus Panel RSV Reagent is used with PathoDx Respiratory Virus Panel.

Specifications
Remel™ PathoDX™ Respiratory Virus Panel RSV Reagent | |
Nasopharyngeal | |
1 x 10 mL |
Respiratory Syncytial Virus (RSV) Antigen | |
Reagent |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Comprehensive Respiratory Virology Testing Solutions
File size: 438.1 kB
PathoDx Respiratory Virus Panel Reagents
Version: FEB.2016
File size: 544.6 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2836552 | Certificate of Analysis | 24/10/2019 | R62411 |
2424071 | Certificate of Analysis | 05/11/2018 | R62411 |
2339432 | Certificate of Analysis | 12/06/2018 | R62411 |
1114008-COA | Certificate of Analysis | 17/04/2017 | R62411 |
973736-COA | Certificate of Analysis | 15/04/2017 | R62411 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel RSV Reagent > TB Potassium Permanganate; 250mL/Bottle
Spot an opportunity for improvement?Share a Content Correction