Learn More
Thermo Scientific™ Remel™ ProSpecT™ Cryptosporidium Sample Panel
Use with ProSpecT Cryptosporidium Microplate Assay, ProSpecT Giardia/Cryptosporidium Microplate Assay, Xpect Cryptosporidium and Xpect Giardia/Cryptosporidium tests.
Brand: Thermo Scientific™ R244803
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Remel Cryptosporidium Sample Panel is used with ProSpecT Cryptosporidium Microplate Assay, ProSpecT Giardia/Cryptosporidium Microplate Assay, Xpect Cryptosporidium and Xpect Giardia/Cryptosporidium tests.
Includes:
5 positive vials and 5 negative vials
Specifications
Cryptosporidium Microplate Assay Sample Panel | |
5 positive vials and 5 negative vials | |
10 Vials/Pk. |
Safety and Handling
- Danger
Hazard Category
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.