Learn More
Thermo Scientific™ Remel™ PYR Disc with Reagent
Use for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Brand: Thermo Scientific™ R30854401
345.31 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ Remel PYR Disk w/Reagent is used for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Specifications
2°C to 8°C | |
Pack of 100 | |
Yes | |
100 Tests |
Safety and Handling
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.