Learn More
Thermo Scientific™ Remel™ Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated)
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated) facilitates cultivation of yeast and fungi.
£80.56
Specifications
Description | Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 |
---|---|
Format | Monoplate - Double + Bag |
Product Type | Agar |
Description
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated) facilitates cultivation of yeast and fungi.
- Settling Plates (Single Bag and Double Bag) are manufactured in a clean room environment
- This provides the highest level of sterility assurance with no compromise of growth characteristics
- Each lot is tested to ensure consistent, reproducible results
-
Specifications
Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 | |
Monoplate - Double + Bag | |
Agar |
Not intended for IVD use.