Learn More
Thermo Scientific™ Remel™ Settling Plate, Sterile, Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar w/Lecithin and Polysorbate 80 facilitates cultivation of yeast and fungi.
Brand: Thermo Scientific™ R111883
Additional Details : Weight : 0.35000kg
Description
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar w/Lecithin and Polysorbate 80 facilitates cultivation of yeast and fungi.
- Double bagged and gamma-irradiated with a third sterile bag enclosed for transporting plates to the laboratory
- Sterile 85mm plates are manufactured in a cleanroom (SAL=106)
- 100mm and 150mm plates are manufactured in an environmentally controlled environment
Specifications
Settling Plate Sterile Sabouraud Dextrose Agar w/Lecithin w/Polysorbate 80 | |
13mm x 85mm Plate - Double + Bag | |
Agar | |
10/Pk. |
Not intended for IVD use.