missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Settling Plate, Sterile, Tryptic Soy Agar (Irradiated)

Product Code. 17205964
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
150mm Plate - Double + Bag
Monoplate - Double + Bag
Unit Size:
Pack of 10
Pack of 20
This item is not returnable. View return policy

Product Code. 17205964

Brand: Thermo Scientific™ R111870

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Sterile Tryptic Soy Agar (Irradiated).

Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Sterile Tryptic Soy Agar (Irradiated). It is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. It is also recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.

  • Settling plates (single bag and double bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
  • Plate size:  85 x 13mm
TRUSTED_SUSTAINABILITY

Specifications

Description Settling Plate Sterile Tryptic Soy Agar
Format Monoplate - Double + Bag
Product Type Agar
Quantity 10/Pk.
Dimensions (Dia. x H) 85 x 13 mm

Not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.