Learn More
Thermo Scientific™ Remel™ SP4 Glucose Broth
Thermo Scientific™ Remel SP4 Glucose Broth (w/thallium acetate, penicillin) is used for isolation of Mycoplasma species.
Brand: Thermo Scientific™ R20376
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ Remel SP4 Glucose Broth (w/thallium acetate, penicillin) is used for isolation of Mycoplasma species.
- Penicillin inhibits gram-positive bacteria
Specifications
SP4 Glucose Broth w/thallium acetate, penicillin | |
D - 5mL | |
Microbiology Media | |
20 x 5 mL |
REF R112585 not intended for IVD use.