missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Staphaurex™ Latex Agglutination Test
Rapidly identify and differentiate staphylococci which possess coagulase and/or protein A, particularly Staphylococcus aureus, from other staphylococci.
£161.00 - £850.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | Positive |
Description | Staphaurex Latex Agglutination Test |
CE Marker | Yes |
Type | Staphylococcus aureus Test Kit |
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
11953712
|
Thermo Scientific™
R30859902 |
Staphaurex Latex Agglutination Test | 400 Tests/Kit | 2°C to 8°C |
£850.00
400 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11494089
|
Thermo Scientific™
R30859901 |
Staphaurex Latex Agglutination Test | 120 Tests/Kit | 2°C to 8°C | N/A | ||||
Description
Rapidly identify and differentiate staphylococci which possess coagulase and/or protein A, particularly Staphylococcus aureus, using the Thermo Scientific™ Staphaurex™ Latex Agglutination Test.
Experience quality and reliability with this rapid and accurate latex test for the identification and differentiation of staphylococcus aureus. White latex particles are coated with human fibrinogen for detection of clumping factor and specific IgG for detection of protein A.When mixed on a slide with a suspension of S. aureus, reaction of clumping factor with the fibrinogen and/or protein A with the IgG causes rapid, strong agglutination of the latex particles.
Proven Performance
- Rely on tried-and-true tests with more than 50 years of combined proven performance
- Maintain confidence with reliable results that have greater than 99% sensitivity and specificity (according to data available in the package inserts)
Workflow Efficiency
- Clear-cut, robust reactions provide easy-to-read results
- Improve productivity with compact kit configurations, color-coded reagents, and convenient workstations
- Flexible use from selective and non-selective media
- Control latex is not required
Economical Solution
- Maximize spending efficiency with the option of purchasing separate kit components
- Save time, money, and resources by turning to one company, one source, for a complete set of Staph identification solutions
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
Staphaurex Latex Agglutination Test | |
Staphylococcus aureus Test Kit | |
Staphylococcus aureus |
Positive | |
Yes | |
Test latex, disposable reaction cards, disposable sampling and mixing sticks |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title