missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptex™ Acid Extraction Kit

Rapidly extract and identify streptococcal Lancefield group antigens from cultured organisms using Streptex™ Acid Extraction Kit.

Brand:  Thermo Scientific™ R30951301


Product Code. 13275789

  • £95.95 / 50 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Rapidly extract and identify streptococcal Lancefield group antigens from cultured organisms using Thermo Scientific™ Remel™ Streptex™ Acid Extraction Kit in conjunction with the Streptex™ Kit (ZL50/R30950501 and ZL61/R30164701). Lancefield showed that group-specific streptococcal antigens can be extracted in soluble form and identified by precipitation reactions with homologous antisera1. The Streptex™ kit requires at least 10 minutes incubation at 37°C to extract the group antigen. Reagents supplied in this kit allow a more rapid extraction of group antigen.

Species of Streptococcus are classified based on their hemolytic properties (alpha and beta). Beta-hemolytic streptococci are further classified by Lancefield grouping. The majority of species of Streptococcus possess group-specific antigens which are usually carbohydrate structural components of the cell wall.

Group-specific antigens are extracted from streptococci by nitrous acid in a simple and short incubation step. The extract is then neutralised and antigens are detected and identified with suspensions of latex particles coated with group-specific antibodies.

  • Ready-to-use: Reagent kit
  • Rapid extraction: Within 1 minute.
  • Ease of identification: Positive result is indicated by aggregation of latex particles.

The acid extraction kit provides reagents for the extraction of Lancefield group A, B, C, F and G at room temperature (18 – 30° C). Extraction is complete within 1 minute. Group D antigen cannot be extracted with these reagents.

The latex suspensions for groups A, B, C, D, F and G are supplied in the Streptex™ Kit (ZL50/R30950501 and ZL61/R30164701) or as individual suspensions.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Streptex and Acid Extraction Kit
beta-haemolytic streptococci
7mL each of Extraction Reagent 1, 2, and 3, disposable tubes, disposable tube holder, mixing sticks, sample dispensers
50 Tests/Kit
Yes
Streptococcus Test Kit
Lancefield A, B, C, D, F and G
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers