Learn More
Thermo Scientific™ Remel™ Streptex™ Extraction Enzyme
Remel Streptex and Extraction Enzyme are used with Streptex kit.
Brand:  Thermo Scientific™ R30951001
39.35 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 13275659
  • £42.80 / 25 tests
Explore more special offers
Description
Description
Streptex™ supplementary reagents are intended for use in the streptococcal grouping system provided by the Streptex kit, which contains all components necessary for the system. These reagents are provided as replacement items. For complete information and correct use of this product, the user is referred to the Instructions for Use which accompanies the Streptex kits (ZL50/R30950501 and ZL61/R30164701).
Ordering Alerts:
Freeze dried.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Streptex and Extraction Enzyme
Streptococcus Test Kit
11 mL/Vial, 25 Tests/Kit
Yes
25 Tests/Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3729830 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog Number
Lot Number3696353 Certificate TypeCertificate of Analysis Date27/10/2023 Catalog Number
Lot Number3696303 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog Number
Lot Number3676684 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog Number
Lot Number3676683 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Remel™ Streptex™ Extraction Enzyme > 11mL/Vial; 25 tests/kit