Learn More
Thermo Scientific™ Streptex™ Rapid Latex Agglutination Test
Isolate, identify and differentiate streptococci Lancefield groups A, B, C, F, and G with acid extraction for fast qualitative detection using the Thermo Scientific™ Streptex™ Rapid Latex Agglutination Test.
Brand: Thermo Scientific™ R30950555
130.02 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Streptex™ Rapid is a rapid latex test system for use in the qualitative detection and identification of the Lancefield group of streptococci. Reagents are provided for groups A, B, C, F and G.
Use this rapid latex agglutination test which provides a complete solution for the isolation and differentiation of Lancefield groups A, B, C, F & G. View distinct, clear results with white latex agglutination on a black background. Individual latex grouping reagents are available as required.With Streptex, group specific antigens are extracted from streptococci in a simple incubation step. Antigens are then identified using polystyrene latex particles which have been coated with group-specific antibodies. These latex particles agglutinate strongly in the presence of homologous antigen and remain in smooth suspension in the absence of homologous antigen.
For a more rapid acid extraction method, Streptex Acid Extraction Kit (Part No. R30951301) is available for use with Streptex. Acid extraction will rapidly extract the Group antigens without the need for incubation. However, Group D antigen is not extracted efficiently with acid extraction; therefore, an alternative method must be used to confirm suspected Group D streptococci.
Proven Performance
- Rely on tried-and-true tests with more than 50 years of combined proven performance
- Maintain confidence with reliable results that have greater than 99% sensitivity (according to data available in the package insert)
- Get easy-to-read results with clear-cut, robust reactions
- Improve productivity with compact kit configurations, color-coded reagents and convenient workstations
- Enjoy flexibility with use from selective and non-selective media—no control latex required
- Maximize spending efficiency with the option of purchasing separate kit components
- Save time, money and resources by turning to one company, one source, for a complete set of Staph identification solutions
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
2°C to 8°C | |
Streptex Rapid Latex Agglutination Test | |
Streptococci (Group A, B, C, F, G) | |
Test latex reagents, extraction reagents, polyvalent positive control, disposable tubes, disposable tube holder, disposable reaction cards, mixing sticks and sample dispensers | |
Lancefield A, B, C, F and G |
Positive | |
Yes | |
Streptococcus Test Kit | |
Polyvalent | |
50 Tests/Kit |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6181905 | Certificate of Analysis | 03/03/2025 | R30950555 |
6132561 | Certificate of Analysis | 17/12/2024 | R30950555 |
6123300 | Certificate of Analysis | 19/10/2024 | R30950555 |
6111000 | Certificate of Analysis | 10/09/2024 | R30950555 |
3813719 | Certificate of Analysis | 04/07/2024 | R30950555 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.