Learn More
Thermo Scientific™ Remel™ Violet Red Bile Glucose Agar (Dehydrated)
Remel Violet Red Bile Glucose Agar (Dehydrated) is used for selective and differential isolation of Enterobacteriaceae
Brand:  Thermo Scientific™ R455302
300.14 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 13426636
  • £264.00 / 500g
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Remel Violet Red Bile Glucose Agar (Dehydrated) is used for selective and differential isolation of Enterobacteriaceae
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Industrial Reference: USP.
Powder
500 g
Violet Red Bile Glucose Agar
Agar
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Not intended for IVD use.