missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Flu A and B Control Swab

Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Flu A and B test kits with Thermo Scientific Xpect Remel Flu A and B Control Swab.

Brand:  Thermo Scientific™ R246003

210.86 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12739270

  • £234.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Flu A and B test kits with Thermo Scientific Xpect Remel Flu A and B Control Swab.

These kits contain positive and negative controls to monitor test performance.

Human influenza A and B viruses cause seasonal epidemics of disease (known as the flu season) almost every winter. Flu A is known to cause flu pandemics. The Influenza A and B test is performed for the qualitative detection and differentiation of Influenza A virus, including H5N1 and H1N1, and Influenza B virus in nasal/throat/nasopharyngeal swab or nasopharyngeal aspirate.

This kit contains Flu A control dry swab containing inactivated influenza A antigen and Flu B control dry swab containing inactivated influenza B antigen.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 8°C
Influenza
20 Flu A+/Flu B- control swabs and 20 Flu A-/Flu B+ control swabs
Xpect Flu A and B Control Swab
Control
40 Swabs
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number970547 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR246003
Lot Number966637 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR246003
Lot Number942860 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR246003
Lot Number920116 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR246003
Lot Number891031 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR246003
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Flu A and B Control Swab > Control Swabs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.