Learn More
Thermo Scientific™ Rotavirus Control Kit
Monitor gross systematic failures using Xpect Rotavirus Control Kit as external quality control material for Xpect Rotavirus Kit.
Brand: Thermo Scientific™ R246551
352.45 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Monitor gross systematic failures using Thermo Scientific™ Remel™ Xpect™ Rotavirus Control Kit as external quality control material for Xpect Rotavirus Kit. This kit contains a negative and a positive control.
Monitor gross systematic failures using Xpect Rotavirus Control Kit as external quality control material for Xpect Rotavirus Kit.
- Ready to Use
- Used as a confirmatory test before the patient sample is used.
- Should be tested according to Xpect Rotavirus Instructions For Use (IFU).
This kit is used for qualitative procedures. The test should produce negative and positive results with the corresponding control before testing the patient sample.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Control Kit | |
1mL/Vial positive control; 1mL/Vial negative control | |
2 Vials/Kit |