Learn More
Thermo Scientific™ Remel™ Xpect™ RSV Control Kit
Remel Xpect RSV Control Kit is an Xpect RSV liquid control set.
Brand: Thermo Scientific™ R246012
470.86 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Remel Xpect RSV Control Kit is an Xpect RSV liquid control set.
- Method: Chromatographic immunoassay
- Samples collected from nasopharyngeal specimens in neonatal and paediatric patients
Ordering Alerts:
Six positive and negative tests can be run from 1 set of controls.
Includes:
1 vial positive control and 1 vial negative control. 1mL/Vial.

Specifications
6 Tests | |
Control | |
1 Vial positive control and 1 Vial negative control. 1mL/Vial. | |
2 Vials |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.