Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for Thermo Scientific™ Instruments
Use Thermo Scientific™ Removable-Needle Syringes for accurate and reproducible sample introduction into GC instruments, with or without the needle.
£63.67 - £151.00
Product Code | Brand | Volume (Metric) | Needle Gauge | Length (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Length (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||||
10702517
|
Thermo Scientific™
36500510 |
5 μL | 31 G | 75 mm | Thermo GCs | N/A | N/A | N/A | |||||
10743867
|
Thermo Scientific™
365D1611 |
10 μL | 23 G | 42 mm | Agilent GC Instruments |
£82.26
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10785174
|
Thermo Scientific™
365D1841 |
10 μL | 26 G | 50 mm | CTC GC Instruments |
£63.67
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10419054
|
Thermo Scientific™
365D3731 |
10 μL | 23 G | 50 mm | CTC GC Instruments |
£63.67
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10724847
|
Thermo Scientific™
36503015 |
50 μL | 23 G | 50 mm | AS2000 Autosampler |
£106.53
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12896036
|
Thermo Scientific™
365B0251 |
1 μL | 23 G | 57 mm | TriPlus RSH Autosampler |
£151.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15274563
|
Thermo Scientific™
365C6610 |
5 μL | 26s G | 42 mm | Shimadzu GC Instruments |
£88.24
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15264573
|
Thermo Scientific™
365D6610 |
10 μL | 26s G | 42 mm | Shimadzu GC Instruments |
£72.96
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Get durability, clarity and accuracy in gas chromatography (GC) sample introduction for confidence in analytical results, time-after-time. Thermo Scientific™ Removable-Needle Syringes offer accurate and reproducible sample introduction, with or without the needle. Choose from a variety of sizes, types and gas chromatography autosampler compatibilities. They are available in standard industry size with black easy-to-read and solvent-resistant markings.
- Manual syringes in industry-standard size
- White scale backing for easy reading
- Solvent-resistant markings
Specifications
Cone |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.