missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Replacement 3.8 Liter Vacuum Aspirator Bottle

Product Code. 15785673
Click to view available options
Capacity (Metric):
3.8 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Replacement bottle for product 15219677

Specifications

Capacity 1.0 Gal (3.8 Liters)
Capacity (Metric) 3.8 L
Capacity (English) 1.0 gal.
Color Blue
Material Polypropylene
No. per Pack 1
Quantity 1
Autoclavable Autoclavable
For Use With (Equipment) Vacuum Aspirator Bottle 15219677
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.