missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Replacement Base Tray for Unstirred Water Baths

Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments Replacement Base Tray for Unstirred Water Baths promote thermal convection in the bath.

Brand:  Grant Instruments™ AQBT12

Code : 57

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11436417

  • £57.00 / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • For use with Sub Aqua Pro Series Unstirred Water Bath
  • Choose a rack that fits 2, 5, 12, or 26L tank size
  • Racks are supplied as standard with purchase of a Sub Aqua Pro Unstirred water Bath
Specifications

Specifications

Sub Aqua Pro 12L
Base Tray
Polycarbonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers