Learn More
Saint-Gobain Replacement Blade
Use this blade with benchtop tubing cutters to cut tubing with an outer diameter of 1/16 to 5/8 in.
Brand: Saint-Gobain 06438-11
Code : A0
Additional Details : Weight : 0.02000kg
Description
- Replacement blade for benchtop tubing cutter
- Steel blade makes right-angle cuts in plastic tubing
Specifications
Replacement Blade |