Learn More
Labconco™ Replacement Connecting Bulbs
These Replacement Connecting Bulbs separate liquid from vapors. Designed for Kjeldahl Units.
Brand: Labconco™ 2081300
Additional Details : Weight : 0.20000kg
Description
- The connecting bulb separates liquid from vapors prior to the vapors entering the condenser.
- Double baffle design effectively traps caustic mist that may carry over to the recovery solution.
- Provide connection between a Kjeldahl flask and distillation rack.
- One-piece borosilicate glass.
- Each piece of glassware may be labeled 20813-01.
Specifications
Connecting Bulb | |
1.8 kg | |
Kjeldahl Units |
Borosilicate Glass | |
4 lb. | |
6 Pk. |