Learn More
Restek™ Replacement FID Collector Assembly for Agilent 6890/6850 GCs
Click to view available options
Type:
FID Collector Assembly Kit
FID Collector Assembly Kit w/Siltek Ignitor Castle
Unit Size:
Each
Description
- Constructed of high-quality stainless steel
- Individual replacement parts available
Specifications
Specifications
| Type | FID Collector Assembly Kit |
| For Use With | Agilent 6890/6850 |