Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
A range of options for secure closure on microcentrifuge rotors to help contain samples.
Brand: Thermo Scientific™ 75003405
98.88 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.02880kg
Description
A range of options for secure closure on microcentrifuge rotors to help contain samples.
- O-rings for inner and outer seals
- ClickSeal lids for optimal biocontainment
- Available for a variety of sizes and models
Specifications
O-ring | |
Rotor Lid | |
Replacement O-ring for MicroClick 24 x 2 Rotor Lid |