Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
A range of options for secure closure on microcentrifuge rotors to help contain samples.
£112.73 - £239.02
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
15145283
|
Thermo Scientific™
75003405 |
O-ring | MicroClick 24 x 2 Rotor Lid |
£112.73
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15165273
|
Thermo Scientific™
75005725 |
ClickSeal Biocontainment Lid | MicroClick 24 x 2 Rotor |
£239.02
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15125263
|
Thermo Scientific™
75005726 |
O-ring | MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors |
£116.37
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15114933
|
Thermo Scientific™
75005730 |
ClickSeal Biocontainment Lid | MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors |
£223.76
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A range of options for secure closure on microcentrifuge rotors to help contain samples.
- O-rings for inner and outer seals
- ClickSeal lids for optimal biocontainment
- Available for a variety of sizes and models
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors