Learn More
Thermo Scientific™ Replacement Microcentrifuge Rotor Lids
Combine these replacement microcentrifuge rotor lids with the microcentrifuge rotor.
Brand:  Thermo Scientific™ 75003406
 View more versions of this product
Product Code. 10554573
  • £308.00 / Each
Explore more special offers
Description
Description
Combine Thermo Scientific™ Replacement Microcentrifuge Rotor Lids with the microcentrifuge rotor.
ClickSeal™ Biocontainment Lid:
  • For use with 24 × 1.5/2.0 mL Rotor (75003424), PCR 4 × 8 Rotor (75003440)
  • Unique design clicks when proper seal is made, providing assurance that your samples are securely contained, as well as shortens retrieval time with one click opening
  • With PPSU rotor lid, view contents of rotor for added safety and convenience
Screw-on Lid:
  • For use with 36 × 0.5 mL Rotor (75003436), Dual Row 18 × 2.00.5 mL Rotor (75003418), PCR 8 × 8 Rotor (75003489)
Ordering Information:
Sold individually.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermo Scientific microcentrifuge rotors 75003436, 75003489, and 75003418
Screw-on Lid