Learn More
Thermo Scientific™ Replacement Needles for GC Syringes
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Brand:  Thermo Scientific™ 36550045
 View more versions of this product
Product Code. 10260095
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
36550045
Replacement Syringe Needle
75 mm
Thermo Scientific 36500510 Syringe
31 G
5 μL
Cone
Removable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Replacement Needles for GC Syringes > For use with Thermo Scientific syringe 36500510
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.