Learn More
Thermo Scientific™ Replacement Needles for LC Syringes
Product Code. 10079142 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
25 to 500 μL
Unit Size:
Pack of 5
Product Code. 10079142
Brand: Thermo Scientific™ 365RNL25
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Replacement Needles for LC Syringes are available for syringes with removable needles.
Thermo Scientific™ Replacement Needles for LC Syringes are available for syringes with removable needles.
TRUSTED_SUSTAINABILITY
Specifications
Description 365RNL25
Needle Gauge 22 G
Length (English) Needle 2 in.
Product Type Syringe Needle
Volume (Metric) 25 to 500 μL
Length (Metric) Needle 50.8 mm
For Use With (Equipment) LC Autosampler Syringe
Connection Type Removable