Learn More
Fisherbrand™ Replacement Parts for MultiSUB Choice Horizontal Gel System
Spare safety lid and electrophoresis tank for use with 15 x 7cm, 15 x 10cm or 15 x 15cm gel trays. Fisherbrand™ Replacement Parts for MultiSUB Choice Horizontal Gel System are constructed of high-quality, injection molded plastic for use in nucleic acid gel electrophoresis applications.
£123.00 - £220.00
Specifications
For Use With (Application) | Nucleic Acid Gel Electrophoresis |
---|---|
For Use With (Equipment) | MultiSUB Choice Horizontal Gel Electrophoresis Unit |
Gel Dimensions | 15 x 7 cm, 15 x 10 cm or 15 x 15 cm |
Material | Molded Plastic |
No. of Wells | For up to 210 separations |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11813303
|
Fisherbrand™
|
Lid |
£123.00
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
11873413
|
Fisherbrand™
|
Tank with Electrodes |
£220.00
Each |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
Description
MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed.
The wider format of the MultiSUB Choice enables the rapid separation of up to 210 samples. A comprehensive selection of combs provides full flexibility for preparatory and screening techniques. MS Choice is suitable for cloning and mini-prep analysis, and it is ideal for restriction fragment analysis, sample prep or checking of high numbers of samples.
- Electrophoresis Tank (Including Electrodes): A raised platform holds the UV sample trays, with side wells to hold buffer solution; connects to an external power source
- Asymmetrical Safety Lid: Notes the correct polarity and orientation of electrodes, with guides that enable the electrode thumb locators to click easily into the correct position
- Tank and lid are designed for use with 15 x 7cm, 15 x 10cm or 15 x 15cm gel trays
Specifications
Nucleic Acid Gel Electrophoresis | |
15 x 7 cm, 15 x 10 cm or 15 x 15 cm | |
For up to 210 separations |
MultiSUB Choice Horizontal Gel Electrophoresis Unit | |
Molded Plastic | |
1 Each |