Learn More
Fisherbrand™ Replacement Parts for MultiSUB Midi Horizontal Gel System
Spare safety lid and electrophoresis tank for use with 10 x 7cm or 10 x 10cm gel trays. Fisherbrand™ Replacement Parts for MultiSUB Midi Horizontal Gel System are constructed of high-quality, injection molded plastic for use in nucleic acid gel electrophoresis applications.
£78.75 - £162.00
Specifications
For Use With (Application) | Nucleic Acid Gel Electrophoresis |
---|---|
For Use With (Equipment) | MultiSUB Midi Horizontal Gel Electrophoresis Unit |
Gel Dimensions | 10 x 7 cm or 10 x 10 cm |
Material | Molded Plastic |
No. of Wells | For up to 100 separations |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11803303
|
Fisherbrand™
FMS10LID |
Lid |
£78.75
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
11853353
|
Fisherbrand™
MS10TANK |
Tank with Electrodes |
£162.00
Each |
Estimated Shipment: 11-11-2024
Log in to see stock. |
|||||
Description
MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed.
The MultiSUB Midi is has a wider format than the Mini and is the preferred option for quick sample checks of medium volumes, particularly during PCR and cloning. It is an economical choice for rapid separation of up to 100 samples.
- Electrophoresis Tank (Including Electrodes): A raised platform holds the UV sample trays, with side wells to hold buffer solution; connects to an external power source
- Asymmetrical Safety Lid: Notes the correct polarity and orientation of electrodes, with guides that enable the electrode thumb locators to click easily into the correct position
- Tank and lid are designed for use with both 10 x 7cm or 10 x 10cm gel trays
Specifications
Nucleic Acid Gel Electrophoresis | |
10 x 7 cm or 10 x 10 cm | |
For up to 100 separations |
MultiSUB Midi Horizontal Gel Electrophoresis Unit | |
Molded Plastic | |
1 Each |