Learn More
Fisherbrand™ Replacement Parts for MultiSUB Mini Horizontal Gel System
Spare safety lid and electrophoresis tank for use with 7 x 7cm or 7 x 10cm gel trays. Fisherbrand™ Replacement Parts for MultiSUB Mini Horizontal Gel System are constructed of high-quality, injection molded plastic for use in nucleic acid gel electrophoresis applications.
£105.00 - £230.00
Specifications
For Use With (Application) | Nucleic Acid Gel Electrophoresis |
---|---|
For Use With (Equipment) | MultiSUB Mini Horizontal Gel Electrophoresis Unit |
Gel Dimensions | 7 x 7 cm or 7 x 10 cm |
Material | Molded Plastic |
No. of Wells | For up to 64 separations |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11833303
|
Fisherbrand™
FMS7LID |
Lid |
£105.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
11827573
|
Fisherbrand™
MS7TANK |
Tank with Electrodes |
£230.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
Description
MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed.
The MultiSUB Mini is the preferred option for quick sample checks of small to medium volumes, particularly following restriction digestion during cloning. It is an economical choice for separation of up to 64 samples.
- Electrophoresis Tank (Including Electrodes): A raised platform holds the UV sample trays, with side wells to hold buffer solution; connects to an external power source
- Asymmetrical Safety Lid: Notes the correct polarity and orientation of electrodes, with guides that enable the electrode thumb locators to click easily into the correct position
- Tank and lid are designed for use with both 7 x 7cm or 7 x 10cm gel trays
Specifications
Nucleic Acid Gel Electrophoresis | |
7 x 7 cm or 7 x 10 cm | |
For up to 64 separations |
MultiSUB Mini Horizontal Gel Electrophoresis Unit | |
Molded Plastic | |
1 Each |