Learn More
Merck Replacement Parts for Stirred Cells
Product Code. p-4376295 Shop All Merck Products
Click to view available options
For Use With (Equipment):
Amicon 8003 Series Stirred Cell
Amicon 8200 Series Stirred Cell
Unit Size:
1 set
Each
Product Code. 11700403
Brand: Merck 8003SCPKIT
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Suitable for use with Amicon™ stirred cell apparatus
  • Small Replacement Parts for 8000 Series Stirred Cells include 2 O-rings (Mfr. Nos. 2122S, 2118S, 2015S), tubing (Mfr. No. 4500202), tubing fitting assembly (Mfr. No. 15025AM, and membrane holder (Mfr. No. 2214801)
  • Large Replacement Parts for Stirred Cells include one each: cap assembly, stirrer assembly and base
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Amicon 8003 Series Stirred Cell
Includes Cap assembly (1524901), stirrer assembly (15247) and base (210000)
Type Replacement Part