Learn More
Grant Instruments™ Polycarbonate Transparent Lid, Gabled
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
Brand:  Grant Instruments™ AQL12
 View more versions of this product
Product Code. 11496417
  • £144.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
  • Polycarbonate construction
  • Clear, gabled lid allows condensation buildup to drip away from immersed vessels.
  • For use with Sub Aqua Pro Series Unstirred Water Baths
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gabled
Model SAP12, JBN12, JBA12 and SBB Aqua 12 Plus SUB Aqua Plus water bath
Used to avoid contamination, reduce evaporation, and save energy
Polycarbonate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Polycarbonate Transparent Lid, Gabled > Capacity: 12L; For 12L Tanks