Learn More
DWK Life Sciences Replacement Seal
The sealing ring serves as an intermediate piece between the plastic olive and the glass thread and enables a leak-proof connection of the components (leakage protection).
Brand: DWK Life Sciences 292200904
5.53 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
A seal is part of a construction and prevents or limits unwanted mass transfer.
- For leak-tight connection of the plastic olives
Specifications
Autoclavable | |
Replacement sealing ring for olive; For plastic hose connections (Cat. No. 29 25506 03 and 29 247 05 04) | |
Silicone (VMQ) |
Plastic Hose Connections (Cat. No. 29 25506 03 and 29 247 05 04) | |
10/Pk. |