missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Color | Product Type | Autoclavable | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Product Type | Autoclavable | Includes | Price | Quantity & Availability | |||
11376390
|
BRAND™
704542 |
Clear, Blue, Red | Pump Assembly | Not Autoclavable | Pump assembly only |
£72.25
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||
12601615
|
BRAND™
704544 |
Clear, Blue, Red | Pump Assembly | Not Autoclavable | Pump assembly only |
£90.35
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||
10705883
|
BRAND™
704549 |
Dark Blue, Red, Translucent Amber | Pump Assembly | Not Autoclavable | Pump assembly only |
£191.00
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||
Description
- 6788 - EPDM seals for seripettor dispensers, pack of 5
- 704536 - autoclavable filling tube for seripettor, 250mm, each
- 704538 - autoclavable filling tube for seripettor, 500mm, each
- 704542 - pump assembly for 1-10mL seripettor
- 704544 - pump assembly for 2.5-25mL seriipettor
- 704549 - Pump assembly for 2.5-25mL seripettor pro
- 704552 - seripettor valve block cap for autoclaving. Fits 1-10mL seripettor
- 704554 seripettor valve block cap for autoclaving. Fits 2.5-10mL seripettor
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Replacement Seals, Pump Assemblies and Valve Block Caps for BRAND seripettor and seripettor pro Bottletop Dispensers