Learn More
Hellma Replacement Silicone Septum Product Code.: 15637115
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
Hellma Replacement Silicone Septum
Product Code. 15637115
10/Pk
Click to view available options
Quantity:
10/Pk
Product Code. 15637115
Brand: Hellma 011651VE1072
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
Add to basket
Firmly fitting and durable silicone seal suitable for a variety of cylindrical, optical cell models.
  • Replacement silicone seal (septum)
  • PTFE-coated on one side
  • Suitable for a variety of cylindrical, optical cell types
Specifications
For Use With (Equipment) Cell models: 117.100; 117.100F, 117.104, 117.104F, 117.200; 117.200F, 117.204, 117.204F
Material Silicone, PTFE
No. per Pack 10/Pk
Quantity 10/Pk
Product Content Correction
Product Title
Hellma Replacement Silicone Septum >