Learn More
Trajan Scientific and Medical™ Replacement SilTite™ Mini Union Ferrules
Click to view available options
For Use With (Equipment):
For 0.1 to 0.25 mm Columns
For 1/32 in. Columns
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
- Specifically designed for connecting capillary columns
- Connects guard columns to capillary columns
- Inert and precise glass lined bore
- Low dead volume
Specifications
Specifications
| Type | SilTite Mini Union Ferrules for 1/32 in. ID Columns |
| For Use With (Equipment) | For 1/32 in. Columns |
| Quantity | 10/Pk. |