Learn More
Trajan Scientific and Medical™ Replacement SilTite™ Mini Union Ferrules
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
£73.75 - £80.80
Product Code | Brand | For Use With (Equipment) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Quantity | Price | Quantity & Availability | ||||
12937111
|
Trajan Scientific and Medical™
073470 |
For 0.1 to 0.25 mm Columns | 10/Pk. |
£80.80
Pack of 10 |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||||
15208933
|
Trajan Scientific and Medical™
073473 |
For 1/32 in. Columns | 10/Pk. |
£73.75
Pack of 10 |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||||
Description
- Specifically designed for connecting capillary columns
- Connects guard columns to capillary columns
- Inert and precise glass lined bore
- Low dead volume
Specifications
10/Pk. |