Learn More
Trajan™ SilTite™ FingerTite Ferrule
For gas chromatography systems
Brand:  Trajan™ 073631
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15228983
  • £70.80 / Pack of 10
Estimated Shipment: 04-10-2024
to see stock.
Add to basket
Description
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
GC Ferrule
10/Pk.
0.5 mm