Learn More
Sartorius™ ReproEasy Performance Test Pads
Test pads for the periodic review of the performance of MA37/160 moisture analyzer.
Brand: Sartorius™ YHP01MA
66.35 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Test pads for the periodic review of the performance of MA37/160 moisture analyzer.
- Allows you to quickly confirm whether a device is working properly and delivers correct results.
- Use together with the performance test function in moisture analyzer.
Specifications
MA37/160 Moisture Analyzer | |
10/Pk. | |
Moisture Analyzer Test Pads, 10/Pk. |