Learn More
Tocris Bioscience™ RepSox
RepSox synthesized to Ancillary Material Grade
Brand: Tocris Bioscience™ TB3742-RMU/10
Additional Details : Weight : 0.00970kg
Description
RepSox is an ancillary Material Grade version of product RepSox (Cat. No. 3742). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.Find out more about our Ancillary Material Grade product range.
Specifications
2-[3-(6-Methylpyridin-2-yl)-1 H-pyrazol-4-yl]-1,5-naphthyridine | |
10 mg | |
C17H13N5 |
446859-33-2 | |
TGF-beta Receptor Inhibitors | |
0.99 |