missing translation for 'onlineSavingsMsg'
Learn More

Resazurin Tablets, Technical, Fisher Chemical™

Product Code. 10395950
Click to view available options
Quantity:
10 Tablets
100 Tablets
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 550-82-3
Molecular Formula C12H7NO4
Molecular Weight (g/mol) 229.191
MDL Number 66739
InChI Key PLXBWHJQWKZRKG-UHFFFAOYSA-N
PubChem CID 11077
ChEBI CHEBI:8806
IUPAC Name 7-hydroxy-10-oxidophenoxazin-10-ium-3-one
SMILES C1=CC2=C(C=C1O)OC3=CC(=O)C=CC3=[N+]2[O-]

Specifications

Packaging Glass Jar
Color Orange to Red
Quantity 10 Tablets
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'