missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ Human Residual DNA Quantitation Kits

Product Code. p-7227346 Shop All Applied Biosystems Products
Click to view available options
Includes:
Assay Kit and Sample Preparation
Assay Kit only
Unit Size:
100 reactions
Each
This item is not returnable. View return policy

Product Code. 18745425

Brand: Applied Biosystems™ A26366

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The resDNASEQ Human Residual DNA Quantitation Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from human cell lines used as expression systems for recombinant proteins.

The resDNASEQ Human Residual DNA Quantitation Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from human cell lines used as expression systems for recombinant proteins. Reliable and rapid, the resDNASEQ system enables sensitive (as low as 15 pg DNA/mL test sample) and specific quantitation of human cell DNA. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types, from in-process samples to final product, whether the sample contains high molecular weight or sheared DNA.

Features of the resDNASEQ Human Residual DNA Quantitation Kit include:
• Highly sensitive quantitation using proven TaqMan real-time qPCR technology
• Consistent performance across the expected range of DNA fragment sizes
• Includes master mix, TaqMan primer/probe mix, and human genomic DNA standard

This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

The resDNASEQ Human Residual DNA Quantitation Kit provides highly sensitive detection of human DNA, allowing you to use small sample volumes to generate accurate results. The broad linear range of TaqMan real-time qPCR technology allows you to test samples with variable levels of human DNA in the same assay, such as in-process samples with higher amounts of DNA or bulk drug substance with very low amounts.

Easy to use
With the resDNASEQ Human Residual DNA Quantitation Kit you perform qPCR analysis to compare the amount of human DNA in your test samples to a standard curve generated with known amounts of purified human genomic DNA standard. The resDNASEQ kit includes almost everything you need to carry out these steps, from master mix to TaqMan primer/probe mix to human genomic DNA standard, as well as step-by-step instructions.

Consistent performance
For accurate quantitation of residual human DNA, assay results must be unaffected by the size of the DNA molecules present in the test sample. With the resDNASEQ system you get consistent assay performance across a range of DNA fragment sizes.

*Due to the high degree of similarity between genomes, DNA from closely-related primate species such as monkey (Vero), chimpanzee, and orangutan are expected be detected by the resDNASEQ Human Quantitation Kit.

Specifications

Content And Storage resDNASEQ Human DNA Control
• Human DNA Control, 30 ng/μL, 40 μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ Human DNA Real-Time PCR Reagents
• Environmental Master Mix (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• Human DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

Detection Method Primer-probe
Format Kit
PCR Method qPCR
Host Cell Human DNA
Label or Dye FAM
Product Line resDNASEQ
Quantity 100 reactions
Shipping Condition Dry Ice
No. of Reactions 100 Reactions
Includes Assay Kit only
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.